Assay Details
| Gene Symbol: Brpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282127.1 | NP_001269056.1 | 11 - 12 | 3396 | 83 | |
| NM_001282128.1 | NP_001269057.1 | 11 - 12 | 3646 | 83 | ||
| GenBank mRNA | AK019522.1 | - | 5 - 6 | 1023 | 83 | |
| BC005647.1 | - | 9 - 10 | 1668 | 83 | ||
| BC138361.1 | - | 11 - 12 | 3220 | 8861381 | 83 | |
| BC145256.1 | - | 11 - 12 | 3470 | 9053809 | 83 | |
| DQ288860.1 | - | 6 - 7 | 2086 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brpf1
Genomic Map