Assay Details
| Gene Symbol: Brpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282126.1 | NP_001269055.1 | 3 - 4 | 2004 | 59 | |
| NM_001282127.1 | NP_001269056.1 | 3 - 4 | 1772 | 59 | ||
| NM_001282128.1 | NP_001269057.1 | 3 - 4 | 2004 | 59 | ||
| NM_030178.2 | NP_084454.1 | 3 - 4 | 1772 | 59 | ||
| GenBank mRNA | AK043372.1 | - | 3 - 4 | 2005 | 59 | |
| AK156375.1 | - | 3 - 4 | 1952 | 59 | ||
| BC005647.1 | - | 1 - 2 | 44 | 59 | ||
| BC031594.1 | - | 1 - 2 | 678 | 59 | ||
| BC046521.1 | - | 3 - 4 | 1712 | 5363697 | 59 | |
| BC138361.1 | - | 3 - 4 | 1596 | 8861381 | 59 | |
| BC145256.1 | - | 3 - 4 | 1828 | 9053809 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brpf1
Genomic Map