Assay Details
| Gene Symbol: Brpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282126.1 | NP_001269055.1 | 6 - 7 | 2424 | 111 | |
| NM_001282127.1 | NP_001269056.1 | 6 - 7 | 2192 | 111 | ||
| NM_030178.2 | NP_084454.1 | 6 - 7 | 2192 | 111 | ||
| GenBank mRNA | AK043372.1 | - | 6 - 7 | 2425 | 111 | |
| AK156375.1 | - | 6 - 7 | 2372 | 111 | ||
| BC005647.1 | - | 4 - 5 | 464 | 111 | ||
| BC046521.1 | - | 6 - 7 | 2132 | 5363697 | 111 | |
| BC138361.1 | - | 6 - 7 | 2016 | 8861381 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brpf1
Genomic Map