Assay Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170673.3 | NP_733773.2 | 13 - 14 | 992 | 108 | |
| GenBank mRNA | AK042692.1 | - | 13 - 14 | 993 | 108 | |
| AK044080.1 | - | 13 - 14 | 958 | 108 | ||
| AK137422.1 | - | 13 - 14 | 993 | 108 | ||
| BC115728.1 | - | 13 - 14 | 942 | 40039295 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpne9
Genomic Map