Assay Details
| Gene Symbol: Cpne9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170673.3 | NP_733773.2 | 20 - 21 | 1643 | 93 | |
| GenBank mRNA | AK042692.1 | - | 20 - 21 | 1644 | 93 | |
| AK044080.1 | - | 20 - 21 | 1609 | 93 | ||
| AK047571.1 | - | 18 - 19 | 2658 | 93 | ||
| AK137422.1 | - | 20 - 21 | 1644 | 93 | ||
| AK220550.1 | - | 12 - 13 | 3007 | 93 | ||
| BC115728.1 | - | 20 - 21 | 1593 | 40039295 | 93 | |
| BQ550677.1 | - | 1 - 2 | 93 | |||
| BQ550678.1 | - | 4 - 5 | 278 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758533 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758532 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758541 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758531 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758538 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758534 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758539 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P147766 | AK220550.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P147766 | BC115728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P147766 | BQ550677.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P147766 | AK047571.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P147766 | AK042692.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P147766 | AK220550.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P147766 | BC115728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P147766 | BQ550677.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P147766 | AK047571.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P147766 | AK042692.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336299 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 51228 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701968 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31162 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908098 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420910 | AK084979.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 154714 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84695 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146223 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109087 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P147766 | AK220550.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P147766 | BC115728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P147766 | BQ550677.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P147766 | AK047571.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P147766 | AK042692.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P147766 | AK220550.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P147766 | BC115728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P147766 | BQ550677.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P147766 | AK047571.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P147766 | AK042692.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758551 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758528 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758553 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758549 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758543 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24758525 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109087 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06030533 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06030555 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146223 | - | - |
| Whole Mouse Genome G4122A | A_51_P147766 | AK220550.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147766 | BC115728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147766 | BQ550677.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147766 | AK047571.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147766 | AK042692.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147766 | AK220550.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147766 | BC115728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147766 | BQ550677.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147766 | AK047571.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P147766 | AK042692.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120026 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109087 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146223 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05247596 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05835851 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05247620 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05099470 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05099458 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05099467 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05835850 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758531 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758539 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758541 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758534 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758533 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758532 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24758538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758556 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24758554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05518051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05453330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354321 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05354339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05758820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05239463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05140443 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758563 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758571 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05453345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05099466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05835851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05054804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05830746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24758613 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10540618 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540619 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540615 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540605 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540601 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540621 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540611 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540604 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540608 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540612 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540609 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540620 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540602 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540603 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540616 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540613 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540600 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540614 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540606 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540607 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540610 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10540617 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454653_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10540599 | - | - |
| Mouse Genome 430 2.0 Array | 1454653_at | - | - |
| Mouse Genome 430 2.0 Array | 1454653_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 211232_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 211232_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600009593.mm.1 | TC0600001244.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600009593.mm.1 | NM_170673.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009593.mm.1 | NM_170673.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009595.mm.1 | TC0600001244.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600009595.mm.1 | NM_170673.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600009595.mm.1 | NM_170673.3 | Refseq |
| Mouse Expression Set 430 | 1454653_at | - | - |
| Mouse Expression Set 430 | 1454653_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454653_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454653_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2760110 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2760110 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101580373 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002760110 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1255211 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1255211 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259738 | - | - |
| mouse-6 v1.1 (Illumina) | scl29637.20_542-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00211232.2_312-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2760110 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101580373 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00211232.2_312-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760110 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101580373 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1255211 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1255211 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259738 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00211232.2_312-S | - | - |

Cpne9
Genomic Map