Assay Details
| Gene Symbol: Camk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133926.2 | NP_598687.1 | 10 - 11 | 1026 | 82 | |
| GenBank mRNA | AK134936.1 | - | 10 - 11 | 1046 | 82 | |
| AK170188.1 | - | 9 - 10 | 1092 | 82 | ||
| AK188088.1 | - | 2 - 3 | 126 | 82 | ||
| AK217859.1 | - | 2 - 3 | 126 | 82 | ||
| BC014825.1 | - | 10 - 11 | 1026 | 3969343 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camk1
Genomic Map