Assay Details
| Gene Symbol: Camk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133926.2 | NP_598687.1 | 4 - 5 | 406 | 94 | |
| GenBank mRNA | AK089742.1 | - | 4 - 5 | 389 | 94 | |
| AK134936.1 | - | 4 - 5 | 426 | 94 | ||
| AK205870.1 | - | 3 - 4 | 250 | 94 | ||
| BC014825.1 | - | 4 - 5 | 406 | 3969343 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Camk1
Genomic Map