Assay Details
| Gene Symbol: Camk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133926.2 | NP_598687.1 | 5 - 6 | 548 | 98 | |
| GenBank mRNA | AK089742.1 | - | 5 - 6 | 531 | 98 | |
| AK134936.1 | - | 5 - 6 | 568 | 98 | ||
| AK170188.1 | - | 4 - 5 | 614 | 98 | ||
| AK179926.1 | - | 1 - 2 | 90 | 98 | ||
| BC014825.1 | - | 5 - 6 | 548 | 3969343 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK170188.1 transcripts.
Set Membership: |
|

Camk1
Genomic Map