Assay Details
| Gene Symbol: Creld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133930.1 | NP_598691.1 | 7 - 8 | 1149 | 69 | |
| GenBank mRNA | AB009020.1 | - | 7 - 8 | 1154 | 69 | |
| AK077726.1 | - | 7 - 8 | 1294 | 69 | ||
| AK155777.1 | - | 7 - 8 | 1159 | 69 | ||
| BC010804.1 | - | 7 - 8 | 1149 | 4239786 | 69 | |
| BC023893.1 | - | 7 - 8 | 1160 | 5310111 | 69 | |
| BC025932.1 | - | 7 - 8 | 1055 | 5102571 | 69 | |
| BC029065.1 | - | 7 - 8 | 1185 | 4935296 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creld1
Genomic Map