Assay Details
| Gene Symbol: Emc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175101.3 | NP_780310.1 | 2 - 3 | 370 | 84 | |
| GenBank mRNA | AK020058.1 | - | 2 - 3 | 341 | 84 | |
| AK075595.1 | - | 2 - 3 | 371 | 84 | ||
| AK167819.1 | - | 2 - 3 | 219 | 84 | ||
| AK206599.1 | - | 2 - 3 | 116 | 84 | ||
| AK213319.1 | - | 2 - 3 | 116 | 84 | ||
| BC004641.1 | - | 2 - 3 | 366 | 3157147 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Emc3
Genomic Map