Assay Details
| Gene Symbol: Emc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175101.3 | NP_780310.1 | 6 - 7 | 734 | 69 | |
| GenBank mRNA | AK075595.1 | - | 6 - 7 | 735 | 69 | |
| AK167819.1 | - | 6 - 7 | 583 | 69 | ||
| AK200024.1 | - | 3 - 4 | 187 | 69 | ||
| BC004641.1 | - | 6 - 7 | 730 | 3157147 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Emc3
Genomic Map