Assay Details
| Gene Symbol: Emc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175101.3 | NP_780310.1 | 7 - 8 | 811 | 63 | |
| GenBank mRNA | AK020058.1 | - | 7 - 8 | 782 | 63 | |
| AK075595.1 | - | 7 - 8 | 812 | 63 | ||
| AK167819.1 | - | 7 - 8 | 660 | 63 | ||
| AK199506.1 | - | 1 - 2 | 80 | 63 | ||
| AK200024.1 | - | 4 - 5 | 264 | 63 | ||
| BC004641.1 | - | 7 - 8 | 807 | 3157147 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760053 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760052 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56658 | BC004641.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56658 | AK167819.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56658 | AK020058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56658 | AK075595.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56658 | NM_175101.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56658 | BC004641.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56658 | AK167819.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56658 | AK020058.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56658 | AK075595.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56658 | NM_175101.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 789611 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 327210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 655585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769123 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 646784 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 89673 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426140 | AK081344.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426140 | BC052153.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426140 | K02582.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426140 | L10076.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426140 | NM_021600.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306268 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186446 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112012 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144775 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117598 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121558 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157471 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56658 | BC004641.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56658 | AK167819.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56658 | AK020058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56658 | AK075595.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56658 | NM_175101.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56658 | BC004641.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56658 | AK167819.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56658 | AK020058.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56658 | AK075595.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56658 | NM_175101.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760036 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24760050 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157471 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06031268 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121558 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06031258 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01421290 | - | - |
| Whole Mouse Genome G4122A | A_52_P56658 | BC004641.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56658 | AK167819.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56658 | AK020058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56658 | AK075595.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56658 | NM_175101.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56658 | BC004641.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56658 | AK167819.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56658 | AK020058.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56658 | AK075595.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56658 | NM_175101.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P989682 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112719 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121558 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01421290 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157471 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05297193 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05297197 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760053 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24760052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24760005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24759953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05297200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760052 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05538957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760032 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760046 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05096262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05371251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05426848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05538960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760048 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760044 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24760009 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10546947 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546950 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546951 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546952 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451145_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10546944 | - | - |
| Mouse Genome 430 2.0 Array | 1451145_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1451145_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66087_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66087_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600022470.mm.1 | TC0600002924.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600022470.mm.1 | NM_175101.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600022470.mm.1 | NM_175101.3 | Refseq |
| Mouse Expression Set 430 | 1451145_PM_s_at | - | - |
| Mouse Expression Set 430 | 1451145_s_at | - | - |
| Murine Genome U74v2 Set | 97862_PM_s_at | - | - |
| Murine Genome U74v2 Set | 97861_at | - | - |
| Murine Genome U74v2 Set | 97861_PM_at | - | - |
| Murine Genome U74v2 Set | 97862_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451145_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451145_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451145_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451145_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 583439 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6770594 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5720441 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6770594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5720441 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005720441 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006770594 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2689083 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228253 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228253 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2689083 | - | - |
| mouse-6 v1.1 (Illumina) | scl28539.8_61-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0001142.1_10-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6770594 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5720441 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28539.8_61-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001142.1_10-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6770594 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5720441 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2689083 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228253 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2859294 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2689083 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2859294 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228253 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28539.8_61-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001142.1_10-S | - | - |

Emc3
Genomic Map