Assay Details
| Gene Symbol: Mtmr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026849.2 | NP_081125.2 | 16 - 17 | 1553 | 73 | |
| GenBank mRNA | AK004345.1 | - | 6 - 7 | 434 | 73 | |
| AK034867.1 | - | 15 - 16 | 1414 | 73 | ||
| AK161445.1 | - | 14 - 15 | 1392 | 73 | ||
| BC012436.1 | - | 14 - 15 | 1081 | 73 | ||
| BC018294.1 | - | 16 - 17 | 1503 | 4021243 | 73 | |
| DV648379.1 | - | 8 - 9 | 612 | 73 | ||
| DV653632.1 | - | 4 - 5 | 297 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr14
Genomic Map