Assay Details
| Gene Symbol: Mtmr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026849.2 | NP_081125.2 | 7 - 8 | 872 | 86 | |
| GenBank mRNA | AK034867.1 | - | 7 - 8 | 819 | 86 | |
| AK161445.1 | - | 5 - 6 | 711 | 86 | ||
| AK207311.1 | - | 4 - 5 | 319 | 86 | ||
| BC018294.1 | - | 7 - 8 | 822 | 4021243 | 86 | |
| CD805346.1 | - | 5 - 6 | 418 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK161445.1 transcripts.
Set Membership: |
|

Mtmr14
Genomic Map