Assay Details
| Gene Symbol: Mtmr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026849.2 | NP_081125.2 | 8 - 9 | 944 | 106 | |
| GenBank mRNA | AK034867.1 | - | 8 - 9 | 891 | 106 | |
| AK161445.1 | - | 6 - 7 | 783 | 106 | ||
| BC012436.1 | - | 6 - 7 | 472 | 106 | ||
| BC018294.1 | - | 8 - 9 | 894 | 4021243 | 106 | |
| CD805346.1 | - | 6 - 7 | 490 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK161445.1 transcripts.
Set Membership: |
|

Mtmr14
Genomic Map