Assay Details
| Gene Symbol: Fxyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173372.1 | NP_001166843.1 | 5 - 6 | 215 | 81 | |
| NM_033648.1 | NP_387468.1 | 5 - 6 | 218 | 81 | ||
| GenBank mRNA | AK018728.1 | - | 5 - 6 | 215 | 81 | |
| AU041511.1 | - | 4 - 5 | 81 | |||
| BC086918.1 | - | 6 - 7 | 479 | 6762406 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fxyd4
Genomic Map