Assay Details
| Gene Symbol: Dguok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162521.1 | NP_001155993.1 | 2 - 3 | 339 | 109 | |
| NM_013764.2 | NP_038792.2 | 2 - 3 | 339 | 109 | ||
| GenBank mRNA | AJ133749.1 | - | 2 - 3 | 264 | 109 | |
| AK035281.1 | - | 2 - 3 | 285 | 109 | ||
| AY037862.1 | - | 2 - 3 | 269 | 109 | ||
| BC012647.1 | - | 2 - 3 | 285 | 108 | ||
| BC094920.1 | - | 2 - 3 | 339 | 30055743 | 109 | |
| BQ746075.1 | - | 2 - 3 | 253 | 109 | ||
| U90524.1 | - | 2 - 3 | 259 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dguok
Genomic Map