Assay Details
| Gene Symbol: Dguok | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162521.1 | NP_001155993.1 | 3 - 4 | 525 | 76 | |
| NM_013764.2 | NP_038792.2 | 3 - 4 | 525 | 76 | ||
| GenBank mRNA | AJ133749.1 | - | 3 - 4 | 450 | 76 | |
| AK035281.1 | - | 3 - 4 | 471 | 76 | ||
| AK167071.1 | - | 2 - 3 | 286 | 76 | ||
| AK185573.1 | - | 1 - 2 | 175 | 76 | ||
| AY037862.1 | - | 3 - 4 | 455 | 76 | ||
| BC012647.1 | - | 3 - 4 | 470 | 76 | ||
| BC094920.1 | - | 3 - 4 | 525 | 30055743 | 76 | |
| BQ746075.1 | - | 3 - 4 | 439 | 76 | ||
| U90524.1 | - | 3 - 4 | 445 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dguok
Genomic Map