Assay Details
| Gene Symbol: Htra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019752.3 | NP_062726.3 | 4 - 5 | 1085 | 83 | |
| GenBank mRNA | AF164513.1 | - | 4 - 5 | 964 | 83 | |
| AF175324.1 | - | 4 - 5 | 981 | 83 | ||
| AK011536.1 | - | 1891 | 83 | |||
| AK146338.1 | - | 4 - 5 | 976 | 83 | ||
| AK159188.1 | - | 4 - 5 | 905 | 83 | ||
| AK215600.1 | - | 2 - 3 | 68 | 83 | ||
| BC033276.1 | - | 4 - 5 | 945 | 83 | ||
| BC049880.1 | - | 4 - 5 | 1085 | 5321861 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Htra2
Genomic Map