Assay Details
| Gene Symbol: Aup1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301649.1 | NP_001288578.1 | 10 - 11 | 1307 | 59 | |
| NM_007517.4 | NP_031543.2 | 11 - 12 | 1458 | 59 | ||
| NR_125891.1 | 11 - 12 | 1520 | 59 | |||
| GenBank mRNA | AK154707.1 | - | 11 - 12 | 1327 | 59 | |
| AK158035.1 | - | 11 - 12 | 1249 | 59 | ||
| AK167335.1 | - | 11 - 12 | 1310 | 59 | ||
| AK171011.1 | - | 11 - 12 | 1261 | 59 | ||
| AK171184.1 | - | 11 - 12 | 1254 | 59 | ||
| AK171202.1 | - | 11 - 12 | 1268 | 59 | ||
| AK171249.1 | - | 11 - 12 | 1248 | 59 | ||
| AK171541.1 | - | 11 - 12 | 1248 | 59 | ||
| AK171621.1 | - | 11 - 12 | 1241 | 59 | ||
| BC016485.1 | - | 11 - 12 | 1251 | 3962982 | 59 | |
| BM900175.1 | - | 1 - 2 | 59 | |||
| U41736.1 | - | 11 - 12 | 1256 | 59 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591146 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591145 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591143 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591142 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591141 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591144 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165984 | BC016485.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165984 | U41736.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165984 | AK171621.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165984 | AK171541.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165984 | BC012699.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165984 | BC016485.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165984 | U41736.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165984 | AK171621.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165984 | AK171541.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P165984 | BC012699.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966958 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 887200 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44129 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26613 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165984 | BC016485.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165984 | U41736.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165984 | AK171621.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165984 | AK171541.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165984 | BC012699.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165984 | BC016485.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165984 | U41736.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165984 | AK171621.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165984 | AK171541.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P165984 | BC012699.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591151 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24591140 | - | - |
| Whole Mouse Genome G4122A | A_51_P165984 | BC016485.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165984 | U41736.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165984 | AK171621.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165984 | AK171541.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165984 | BC012699.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165984 | BC016485.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165984 | U41736.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165984 | AK171621.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165984 | AK171541.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P165984 | BC012699.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138973 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591142 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591146 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591140 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591141 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591143 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591144 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24591145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24591194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24591154 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10539286 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539284 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539292 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539280 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539281 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539293 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539282 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539283 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539285 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10539287 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1415742_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10539279 | - | - |
| Mouse Genome 430 2.0 Array | 1415742_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1415742_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11993_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11993_at | - | - |
| Mouse Expression Set 430 | 1415742_PM_at | - | - |
| Mouse Expression Set 430 | 1415742_at | - | - |
| Mouse Expression Set 430 | 1415742_at | - | - |
| Mouse Expression Set 430 | 1415742_PM_at | - | - |
| Murine Genome U74v2 Set | 93809_PM_at | - | - |
| Murine Genome U74v2 Set | 93809_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415742_at | - | - |
| Mouse Genome 430A 2.0 Array | 1415742_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415742_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1415742_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415742_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1415742_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4150075 | - | - |
| MouseWG-6 v2 Expression BeadChip | 101940138 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4150075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6980551 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 101940138 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004150075 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006980551 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2682623 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252199 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252199 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2682623 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2448176 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259485 | - | - |
| mouse-6 v1.1 (Illumina) | scl000066.1_125-S | - | - |
| mouse-6 v1.1 (Illumina) | scl29854.5.1_26-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000070.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 101940138 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6980551 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4150075 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl29854.5.1_26-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000070.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4150075 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980551 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101940138 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2682623 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1252199 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1252199 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2682623 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1259485 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl29854.5.1_26-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000070.1_2-S | - | - |

Aup1
Genomic Map