Assay Details
| Gene Symbol: Aup1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301649.1 | NP_001288578.1 | 6 - 7 | 854 | 57 | |
| NM_007517.4 | NP_031543.2 | 7 - 8 | 1005 | 57 | ||
| NR_125891.1 | 7 - 8 | 1005 | 57 | |||
| GenBank mRNA | AK154707.1 | - | 7 - 8 | 874 | 57 | |
| AK158035.1 | - | 7 - 8 | 796 | 57 | ||
| AK167335.1 | - | 7 - 8 | 795 | 57 | ||
| AK171011.1 | - | 7 - 8 | 808 | 57 | ||
| AK171184.1 | - | 7 - 8 | 801 | 57 | ||
| AK171202.1 | - | 7 - 8 | 815 | 57 | ||
| AK171249.1 | - | 7 - 8 | 795 | 57 | ||
| AK171541.1 | - | 7 - 8 | 795 | 57 | ||
| AK171621.1 | - | 7 - 8 | 788 | 57 | ||
| AK208846.1 | - | 1 - 2 | 65 | 57 | ||
| BC012699.1 | - | 6 - 7 | 638 | 3986517 | 57 | |
| BC016485.1 | - | 7 - 8 | 798 | 3962982 | 57 | |
| U41736.1 | - | 7 - 8 | 803 | 57 | ||
More Information
Set Membership: |
|

Aup1
Genomic Map