Assay Details
| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009357.2 | NP_033383.1 | 1 - 2 | 165 | 66 | |
| GenBank mRNA | AK149789.1 | - | 1 - 2 | 130 | 66 | |
| AK157834.1 | - | 1 - 2 | 144 | 66 | ||
| AK201329.1 | - | 1 - 2 | 62 | 66 | ||
| BC003802.1 | - | 1 - 2 | 109 | 3486968 | 66 | |
| X81058.1 | - | 1 - 2 | 111 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tex261
Genomic Map