Assay Details
| Gene Symbol: Plagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142052.1 | - | 2 - 3 | 224 | 59 | |
| AK183830.1 | - | 1 - 2 | 41 | 59 | ||
| X95503.1 | - | 2 - 3 | 148 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK142052.1, X95503.1 transcripts.
Set Membership: |
|

Plagl1
Genomic Map