Assay Details
| Gene Symbol: Plagl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | X95503.1 | - | 3 - 4 | 195 | 153 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the X95503.1 transcripts.
Set Membership: |
|

Plagl1
Genomic Map