Assay Details
| Gene Symbol: Atn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007881.4 | NP_031907.2 | 2 - 3 | 370 | 101 | |
| GenBank mRNA | BC050920.1 | - | 2 - 3 | 370 | 6406687 | 101 |
| BC053051.1 | - | 2 - 3 | 198 | 5706586 | 101 | |
| D87744.1 | - | 1 - 2 | 80 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC050920.1, BC053051.1, D87744.1, NM_007881.4 transcripts.
Set Membership: |
|

Atn1
Genomic Map