Assay Details
| Gene Symbol: Atn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007881.4 | NP_031907.2 | 3 - 4 | 506 | 84 | |
| GenBank mRNA | BC050920.1 | - | 3 - 4 | 506 | 6406687 | 84 |
| BC053051.1 | - | 3 - 4 | 334 | 5706586 | 84 | |
| D87744.1 | - | 2 - 3 | 216 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atn1
Genomic Map