Assay Details
| Gene Symbol: P3h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013534.4 | NP_038562.2 | 12 - 13 | 1814 | 92 | |
| GenBank mRNA | AJ441086.1 | - | 12 - 13 | 1814 | 92 | |
| AK140962.1 | - | 11 - 12 | 2048 | 92 | ||
| AY463530.1 | - | 12 - 13 | 1814 | 92 | ||
| BC003726.1 | - | 12 - 13 | 1400 | 3497610 | 92 | |
| BC016431.1 | - | 11 - 12 | 1300 | 3483280 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

P3h3
Genomic Map