Assay Details
| Gene Symbol: Cd163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053094.2 | NP_444324.2 | 15 - 16 | 3411 | 102 | |
| GenBank mRNA | AF274883.1 | - | 15 - 16 | 3406 | 102 | |
| BC145793.1 | - | 15 - 16 | 3362 | 40131096 | 102 | |
| DQ058616.1 | - | 15 - 16 | 3367 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd163
Genomic Map