Assay Details
| Gene Symbol: Gnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013530.1 | NP_038558.1 | 8 - 9 | 1081 | 85 | |
| GenBank mRNA | AK014789.1 | - | 8 - 9 | 800 | 85 | |
| AK030354.1 | - | 7 - 8 | 1022 | 85 | ||
| BC018239.1 | - | 8 - 9 | 1081 | 4511683 | 85 | |
| U38494.1 | - | 1 - 2 | 103 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnb3
Genomic Map