Assay Details
| Gene Symbol: Lpcat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145130.2 | NP_660112.1 | 9 - 10 | 1147 | 112 | |
| GenBank mRNA | AB294194.1 | - | 9 - 10 | 1042 | 112 | |
| AK083687.1 | - | 7 - 8 | 703 | 112 | ||
| AK098090.1 | - | 9 - 10 | 1118 | 112 | ||
| AK154062.1 | - | 9 - 10 | 1141 | 112 | ||
| AK165319.1 | - | 9 - 10 | 1126 | 112 | ||
| AK169452.1 | - | 8 - 9 | 1264 | 112 | ||
| AY028317.1 | - | 9 - 10 | 1123 | 112 | ||
| BC006753.1 | - | 9 - 10 | 1075 | 3709076 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lpcat3
Genomic Map