Assay Details
| Gene Symbol: Nop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138747.2 | NP_620086.2 | 4 - 5 | 374 | 67 | |
| GenBank mRNA | AK079914.1 | - | 4 - 5 | 348 | 67 | |
| AK154006.1 | - | 4 - 5 | 328 | 67 | ||
| AK156568.1 | - | 4 - 5 | 321 | 67 | ||
| AK185452.1 | - | 1 - 2 | 98 | 67 | ||
| AK192869.1 | - | 1 - 2 | 66 | 67 | ||
| BY244994.1 | - | 4 - 5 | 374 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nop2
Genomic Map