Assay Details
| Gene Symbol: Nop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK154006.1 | - | 6 - 7 | 565 | 87 | |
| AK156568.1 | - | 6 - 7 | 558 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Nop2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK154006.1 | - | 6 - 7 | 565 | 87 | |
| AK156568.1 | - | 6 - 7 | 558 | 87 | ||
Set Membership: |
|