Assay Details
| Gene Symbol: Clstn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153508.4 | NP_705728.1 | 2 - 3 | 490 | 91 | |
| GenBank mRNA | AJ278486.1 | - | 2 - 3 | 357 | 91 | |
| AK032336.1 | - | 2 - 3 | 432 | 91 | ||
| BC041781.1 | - | 2 - 3 | 189 | 91 | ||
| BC055054.1 | - | 2 - 3 | 422 | 6493096 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Clstn3
Genomic Map