Assay Details
| Gene Symbol: Dhdds | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004993.1 | - | 1 - 2 | 141 | 131 | |
| AK014317.1 | - | 1 - 2 | 141 | 131 | ||
| AK017556.1 | - | 1 - 2 | 152 | 131 | ||
| AK034277.1 | - | 1 - 2 | 141 | 131 | ||
| AK036929.1 | - | 1 - 2 | 157 | 131 | ||
| AK135087.1 | - | 1 - 2 | 141 | 131 | ||
| AK149138.1 | - | 1 - 2 | 111 | 131 | ||
| AK171207.1 | - | 1 - 2 | 152 | 132 | ||
| BC004011.1 | - | 1 - 2 | 127 | 3493585 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034277.1, AK036929.1, AK135087.1, AK149138.1, AK171207.1, BC004011.1 transcripts.
Set Membership: |
|

Dhdds
Genomic Map