| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400012963.mm.1 |
NM_001039048.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012963.mm.1 |
NM_001039048.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012963.mm.1 |
TC0400001631.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400012961.mm.1 |
NM_001039048.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012961.mm.1 |
NM_001039048.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012961.mm.1 |
TC0400001631.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400012960.mm.1 |
NM_001039048.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012960.mm.1 |
NM_001039048.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012960.mm.1 |
TC0400001631.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400012964.mm.1 |
NM_001039048.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012964.mm.1 |
NM_001039048.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400012964.mm.1 |
TC0400001631.mm.1 |
MTA TC |