Assay Details
| Gene Symbol: Trim63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC147663.1 | - | 5 - 6 | 903 | 9087452 | 123 |
| BC147666.1 | - | 5 - 6 | 903 | 9087455 | 123 | |
More Information
Set Membership: |
|

Trim63
Genomic Map