| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400030874.mm.1 |
TC0400003747.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030874.mm.1 |
NM_025382.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030874.mm.1 |
NM_025382.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030882.mm.1 |
TC0400003747.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030882.mm.1 |
NM_025382.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030882.mm.1 |
NM_025382.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030881.mm.1 |
TC0400003747.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030881.mm.1 |
NM_025382.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030881.mm.1 |
NM_025382.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030879.mm.1 |
TC0400003747.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030879.mm.1 |
NM_025382.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030879.mm.1 |
NM_025382.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030883.mm.1 |
TC0400003747.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030883.mm.1 |
NM_025382.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030883.mm.1 |
NM_025382.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030878.mm.1 |
TC0400003747.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030878.mm.1 |
NM_025382.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030878.mm.1 |
NM_025382.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030880.mm.1 |
TC0400003747.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030880.mm.1 |
NM_025382.6.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030880.mm.1 |
NM_025382.6 |
Refseq |