Assay Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025382.6 | NP_079658.2 | 4 - 5 | 705 | 66 | |
| GenBank mRNA | AK038972.1 | - | 5 - 6 | 867 | 66 | |
| AY364165.1 | - | 4 - 5 | 490 | 66 | ||
| AY846873.1 | - | 4 - 5 | 706 | 66 | ||
| AY846874.1 | - | 4 - 5 | 706 | 66 | ||
| BC019186.1 | - | 4 - 5 | 674 | 66 | ||
| BC042733.2 | - | 4 - 5 | 645 | 66 | ||
| BC043306.1 | - | 1 - 2 | 137 | 66 | ||
| BC044802.1 | - | 4 - 5 | 645 | 66 | ||
| BC054769.1 | - | 4 - 5 | 598 | 6415086 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem57
Genomic Map