Assay Details
| Gene Symbol: Ubxn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026257.3 | NP_080533.1 | 10 - 11 | 948 | 75 | |
| GenBank mRNA | AK010492.1 | - | 3 - 4 | 427 | 75 | |
| AK015720.1 | - | 10 - 11 | 932 | 75 | ||
| AK048494.1 | - | 10 - 11 | 950 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ubxn11
Genomic Map