Assay Details
| Gene Symbol: Ubxn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026257.3 | NP_080533.1 | 11 - 12 | 1065 | 69 | |
| GenBank mRNA | AK010492.1 | - | 4 - 5 | 544 | 69 | |
| AK015720.1 | - | 11 - 12 | 1049 | 69 | ||
| AK048494.1 | - | 11 - 12 | 1067 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ubxn11
Genomic Map