Assay Details
| Gene Symbol: Aim1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162970.1 | NP_001156442.1 | 10 - 11 | 3450 | 125 | |
| GenBank mRNA | AK145690.1 | - | 10 - 11 | 3456 | 125 | |
| BC042709.1 | - | 5 - 6 | 471 | 125 | ||
| BC150738.1 | - | 9 - 10 | 3379 | 9087649 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aim1l
Genomic Map