Assay Details
| Gene Symbol: Aim1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162970.1 | NP_001156442.1 | 12 - 13 | 3713 | 71 | |
| GenBank mRNA | AK145690.1 | - | 12 - 13 | 3719 | 71 | |
| BC042709.1 | - | 7 - 8 | 734 | 71 | ||
| BC150738.1 | - | 11 - 12 | 3642 | 9087649 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aim1l
Genomic Map