Assay Details
| Gene Symbol: Cnksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081047.1 | NP_001074516.1 | 13 - 14 | 1205 | 85 | |
| GenBank mRNA | AK145910.1 | - | 13 - 14 | 1167 | 85 | |
| BC113805.1 | - | 13 - 14 | 1170 | 40062208 | 85 | |
| BC113956.1 | - | 13 - 14 | 1170 | 40062209 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnksr1
Genomic Map