Assay Details
| Gene Symbol: Cnksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081047.1 | NP_001074516.1 | 6 - 7 | 715 | 96 | |
| GenBank mRNA | AK145910.1 | - | 6 - 7 | 677 | 96 | |
| BC113805.1 | - | 6 - 7 | 680 | 40062208 | 96 | |
| BC113956.1 | - | 6 - 7 | 680 | 40062209 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnksr1
Genomic Map