Assay Details
| Gene Symbol: Slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039677.2 | NP_001034766.1 | 5 - 6 | 967 | 80 | |
| GenBank mRNA | AK031425.1 | - | 5 - 6 | 763 | 80 | |
| BC013478.1 | - | 4 - 5 | 987 | 80 | ||
| BC113751.1 | - | 5 - 6 | 967 | 40057211 | 80 | |
| BC147704.1 | - | 5 - 6 | 798 | 9087463 | 80 | |
| BC147707.1 | - | 5 - 6 | 798 | 9087466 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a2
Genomic Map