Assay Details
| Gene Symbol: Slc30a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039677.2 | NP_001034766.1 | 7 - 8 | 1214 | 53 | |
| GenBank mRNA | AK031425.1 | - | 7 - 8 | 1011 | 53 | |
| BC013478.1 | - | 6 - 7 | 1234 | 53 | ||
| BC113751.1 | - | 7 - 8 | 1214 | 40057211 | 53 | |
| BC147704.1 | - | 7 - 8 | 1045 | 9087463 | 53 | |
| BC147707.1 | - | 7 - 8 | 1045 | 9087466 | 53 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23322291 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23322288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23322289 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23322292 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23322287 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016732 | BC147704.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2016732 | BC147707.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11692 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 139199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 49718 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2401 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27719 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016732 | BC147704.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2016732 | BC147707.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322303 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322294 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322292 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322299 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322295 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322288 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322296 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322298 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322302 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23322278 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05411715 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05411684 | - | - |
| Whole Mouse Genome G4122A | A_51_P306287 | AK031425.1 | mouse_transcriptome |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23322288 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23322289 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23322291 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23322287 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23322292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322305 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23322385 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03673060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322329 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03671678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04084452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03264397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03831434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322300 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03279086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03210397 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03210399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03671674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322260 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322293 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322312 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03693351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03278767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03269568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322318 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322305 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03511122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03671677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03693354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322299 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322278 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03210400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03511121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322308 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03886587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03671679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03413757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23322304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04084453 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03264396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03210387 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10508968 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508965 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508962 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508964 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508969 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508971 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508966 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508967 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508963 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508970 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427339_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10508961 | - | - |
| Mouse Genome 430 2.0 Array | 1427339_at | - | - |
| Mouse Genome 430 2.0 Array | 1427339_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400012991.mm.1 | NM_001039677.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012991.mm.1 | NM_001039677.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012991.mm.1 | TC0400001633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400012994.mm.1 | NM_001039677.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012994.mm.1 | NM_001039677.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012994.mm.1 | TC0400001633.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400012995.mm.1 | NM_001039677.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012995.mm.1 | NM_001039677.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012995.mm.1 | TC0400001633.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427339_PM_at | - | - |
| Mouse Expression Set 430 | 1427339_at | - | - |
| Murine Genome U74v2 Set | 133053_at | - | - |
| Murine Genome U74v2 Set | 133053_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427339_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427339_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427339_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427339_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 711471 | - | - |

Slc30a2
Genomic Map