Assay Details
| Gene Symbol: Aim1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162970.1 | NP_001156442.1 | 17 - 18 | 4262 | 55 | |
| GenBank mRNA | AK145690.1 | - | 17 - 18 | 4269 | 55 | |
| BC042709.1 | - | 12 - 13 | 1283 | 55 | ||
| BC150738.1 | - | 16 - 17 | 4191 | 9087649 | 55 | |
| BP759371.1 | - | 3 - 4 | 55 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Aim1l
Genomic Map