Assay Details
| Gene Symbol: Aim1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162970.1 | NP_001156442.1 | 19 - 20 | 4578 | 65 | |
| GenBank mRNA | AK145690.1 | - | 19 - 20 | 4585 | 65 | |
| AK165037.1 | - | 5 - 6 | 2539 | 65 | ||
| BC042709.1 | - | 14 - 15 | 1599 | 65 | ||
| BC077737.1 | - | 2 - 3 | 585 | 65 | ||
| BC150738.1 | - | 18 - 19 | 4507 | 9087649 | 65 | |
| BP759371.1 | - | 1 - 2 | 65 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019300 | BC150738.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019300 | NM_001162970.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2019300 | AK145690.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564036 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290129 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894039 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 228532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853537 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264651 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423309 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739357 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 836232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 170093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 133127 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 56331 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154971 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149421 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019300 | BC150738.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019300 | NM_001162970.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2019300 | AK145690.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00994170 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154971 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149421 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05410990 | - | - |
| Whole Mouse Genome G4122A | A_52_P559066 | BP759371.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559066 | BC077737.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559066 | BC150738.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559066 | AK165037.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P559066 | AK155034.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149421 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03918623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03530301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04098924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320910 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03819856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03848259 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03587957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03587954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03611578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03673761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936434 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03587952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03416446 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936457 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03423590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03205684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04098934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03673756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03423589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03514533 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03807210 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03587961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03760286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03587960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320919 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03559913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03760289 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03819847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03611581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320817 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03416448 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03981500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03423591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03587963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03230777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03848258 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03230779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03819855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03230784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03230774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320826 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03530288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03819852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03787041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320795 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03819849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03834127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320859 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03819853 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23320848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03936441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03787040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03587962 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10508933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508932 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508921 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508929 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10508924 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437813_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10508917 | - | - |
| Mouse Genome 430 2.0 Array | 1437813_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437813_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230806_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 230806_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400012876.mm.1 | NM_001162970.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012876.mm.1 | NM_001162970.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012876.mm.1 | TC0400001626.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400012878.mm.1 | NM_001162970.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012878.mm.1 | NM_001162970.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012878.mm.1 | TC0400001626.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400012877.mm.1 | NM_001162970.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012877.mm.1 | NM_001162970.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400012877.mm.1 | TC0400001626.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437813_at | - | - |
| Mouse Expression Set 430 | 1437813_PM_at | - | - |
| Murine Genome U74v2 Set | 168165_at | - | - |
| Murine Genome U74v2 Set | 168165_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437813_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437813_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 777124 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100050746 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1214944 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224946 | - | - |

Aim1l
Genomic Map