Assay Details
| Gene Symbol: Tbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011603.5 | NP_035733.1 | 5 - 6 | 763 | 77 | |
| GenBank mRNA | AB017697.1 | - | 5 - 6 | 660 | 77 | |
| AK005604.1 | - | 5 - 6 | 582 | 76 | ||
| AK049975.1 | - | 5 - 6 | 635 | 77 | ||
| AK078547.1 | - | 5 - 6 | 695 | 77 | ||
| BC033926.1 | - | 5 - 6 | 604 | 1382180 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbpl1
Genomic Map