Assay Details
| Gene Symbol: Tbpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011603.5 | NP_035733.1 | 6 - 7 | 857 | 99 | |
| GenBank mRNA | AB017697.1 | - | 6 - 7 | 754 | 99 | |
| AK005604.1 | - | 6 - 7 | 676 | 99 | ||
| AK049975.1 | - | 6 - 7 | 729 | 99 | ||
| AK078547.1 | - | 6 - 7 | 789 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422629 | BC033926.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422629 | AK005604.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422629 | AK049975.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422629 | AK078547.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422629 | NM_011603.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422629 | BC033926.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422629 | AK005604.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422629 | AK049975.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422629 | AK078547.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P422629 | NM_011603.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2018442 | BC033926.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 418714 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 888919 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 144577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945085 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322226 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390573 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966346 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609476 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193649 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 57853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 29075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 71677 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153652 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102279 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2018442 | BC033926.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422629 | BC033926.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422629 | AK005604.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422629 | AK049975.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422629 | AK078547.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422629 | NM_011603.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422629 | BC033926.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422629 | AK005604.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422629 | AK049975.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422629 | AK078547.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P422629 | NM_011603.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422629 | BC033926.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422629 | AK005604.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422629 | AK049975.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422629 | AK078547.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422629 | NM_011603.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422629 | BC033926.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422629 | AK005604.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422629 | AK049975.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422629 | AK078547.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422629 | NM_011603.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P64924 | BE650743.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P64924 | AK049975.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P64924 | AK078547.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P64924 | NM_011603.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422636 | BC033926.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422636 | AK005604.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422636 | AK049975.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422636 | AK078547.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P422636 | NM_011603.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422636 | BC033926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422636 | AK005604.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422636 | AK049975.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422636 | AK078547.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422636 | NM_011603.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P387690 | - | - |
| Whole Mouse Genome G4122A | A_51_P422629 | BC033926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422629 | AK005604.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422629 | AK049975.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422629 | AK078547.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422629 | NM_011603.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422629 | BC033926.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422629 | AK005604.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422629 | AK049975.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422629 | AK078547.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P422629 | NM_011603.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P64924 | BE650743.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P64924 | AK049975.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P64924 | AK078547.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P64924 | NM_011603.5 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115052 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127820 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08645261 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08950405 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08645277 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08950407 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08645263 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08645264 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10368237 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368239 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368238 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368232 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368233 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10368236 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434907_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431415_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10368229 | - | - |
| Mouse Genome 430 2.0 Array | 1431415_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1434907_at | - | - |
| Mouse Genome 430 2.0 Array | 1434907_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431415_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237336_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 237336_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000013448.mm.1 | NM_011603.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013448.mm.1 | NM_011603.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013448.mm.1 | TC1000001863.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000013447.mm.1 | NM_011603.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013447.mm.1 | NM_011603.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013447.mm.1 | TC1000001863.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000013445.mm.1 | NM_011603.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013445.mm.1 | NM_011603.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000013445.mm.1 | TC1000001863.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431415_PM_a_at | - | - |
| Mouse Expression Set 430 | 1431415_a_at | - | - |
| Mouse Expression Set 430 | 1434907_at | - | - |
| Mouse Expression Set 430 | 1434907_PM_at | - | - |
| Murine Genome U74v2 Set | 97893_PM_at | - | - |
| Murine Genome U74v2 Set | 97893_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431415_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1431415_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431415_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1431415_a_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434907_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434907_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 353530 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3610332 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3610332 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003610332 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2605872 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2605872 | - | - |
| mouse-6 v1.1 (Illumina) | scl38128.9_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3610332 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38128.9_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3610332 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2605872 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2605872 | - | - |

Tbpl1
Genomic Map